missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Legionella BCYE Supplement w/o Cysteine
Description
Oxoid Legionella BCYE Supplement w/o L-cysteine is used for the isolation of Legionella spp.
- Add to Legionella CYE Agar Base, Part No. CM1203B
- Each vial supplements 100mL of medium
Specifications
Specifications
| Description | Legionella BCYE Supplement w/o L-cysteine |
| Form | Freeze-dried |
| Product Type | Microbiology Media |
| Quantity | 10/Pk. |
| Yield | Each vial for 100mL medium |
| Species | Legionella |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?