missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Desoxycholate Citrate Agar (Dehydrated)

Product Code. 10076012
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10076012 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10076012 Supplier Thermo Scientific™ Supplier No. CM0035B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate and recover intestinal pathogens using this medium.

Isolate and recover intestinal pathogens using Thermo Scientific™ Oxoid™ Desoxycholate Citrate Agar (Dehydrated).

The medium is pale pink and clear. Lactose fermenting organisms produce pink colonies and may be surrounded by a zone of precipitated desoxycholic acid. The colonies of non-lactose fermenters are colorless, and due to their alkaline reaction, they are surrounded by a clear orange-yellow zone of medium.

Desoxycholate Citrate Agar is a modification of Leifson medium1, for the isolation and maximum recovery of intestinal pathogens. It is less selective and inhibiting than Desoxycholate Citrate Agar (Hynes) CM0227.

  • Easy-to-read: provides an opaque background to easily discern the clearing produced by alkali-producing pathogens.
  • Preferable when shigellae are being sought as well as salmonellae2.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Desoxycholate Citrate Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 10.3L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.