missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Crossley Milk Medium (Dehydrated)

Product Code. 10107612
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10107612 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10107612 Supplier Thermo Scientific™ Supplier No. CM0213B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carry out routine examination of canned food samples for anaerobes with this medium.

Carry out routine examination of canned food samples for anaerobes with Thermo Scientific™ Oxoid™ Crossley Milk Medium (Dehydrated). The medium gives rapid growth without the use of special anaerobic apparatus while allowing the detected bacteria to be provisionally identified by their reactions upon the medium.

Canned foods are more often prone to flat sour spoilage due to contamination by either mesophilic or thermophilic anaerobic spore formers.
Crossley milk medium is recommended, in the second edition of Tanner’s `The Microbiology of Foods’1, for the examination of meat, meat products, and canned foods for sporing anaerobes.

Organism Diagnostic reaction Organism indicated
 1 Neutral or alkaline pH (purple colour), gas production,soft curd followed by rapid digestion of casein, often to clear brown liquid, formation of black sediment accompanied by typical foul odor Cl. putrificum
Cl. sporogenes
Cl. flabelliferum
Cl oedematiens
Cl. histolyticum
 2 No initial change of pH, formation of soft curd within 2-3 days, slight gas formation. Complete digestion later with alkaline reaction, no odor Cl. centrosporogenes
 3 Slight acidity (pale yellow colour), formation of soft curd, and whey. Slight gas production Cl. sphenoides
 4 Acid (bright yellow colour), formation of firm clot and gas. Bleaching of the indicator may sometimes occur Cl. butyricum
 5 (a) Acid, formation of ’stormy’ clot.
(b) Acid and ’stormy’ clot but with less gas and cloudy whey
(a) Cl. perfingens
(b) Usually Cl. tertium
 6 Strong alkaline pH, with peptonization commencing at the surface and spreading downwards. Digestion not complete, no blackening, no odor, no gas production B. subtilis
B. vulgaris
 7 Acid and clot, or slightly acid only. Peptonization in some cases B. cereus
B. coagulans
B. silvaticus
and various cocci (more detailed tests required)

Crossley2 modified his medium, by the addition of 20% (w/v) of autoclaved meat or fish paste, so that it was suitable for the examination of vegetable and dairy products.
The medium recommended by Jepsen3 in `Meat Hygiene’ published by the World Health Organization, is Crossley Milk Medium modified by the addition of 20% (w/v) of cooked fish; it is suitable for the examination of meat products for clostridia, and gives diagnostic reactions essentially similar to those outlined above. Riemann4 modified Crossley Milk Medium by the addition of 0.08% of cysteine hydrochloride, before autoclaving, or by the addition of 1mL of a sterile 10% sodium thioglycollate solution just before use.

Not all products are available for sale in all territories. Please inquire.
 
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand


TRUSTED_SUSTAINABILITY

Specifications

Description Crossley Milk Medium
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 4.5L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.