missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Baird-Parker RPF Agar Base

Product Code. 10026452
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10026452 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10026452 Supplier Thermo Scientific™ Supplier No. CM0961B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective medium for the isolation and enumeration of coagulase positive staphylococci from food.

The Oxoid Baird-Parker RPF Agar Base is a selective medium for the isolation and enumeration of coagulase positive staphylococci from food.

  • For use with RPF supplement, Part No. SR0122A
  • Do not use Egg Yolk Tellurite Emulsion
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: ISO, NMKL.
Description Baird-Parker Agar Base, RPF
Form Powder
Product Type Agar
Quantity 500 g
Yield For 7.1L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.