Learn More
Thermo Scientific™ Owl™ Blotting Paper
Absorb adequate amounts of buffer for semi-dry electroblotting without the loss of mechanical stability with this high-quality blotting paper.
Brand: Thermo Scientific™ FP-4
39.81 GBP valid until 2024-12-31
Use promo code "21973" to get your promotional price.
Code : 04
Additional Details : Weight : 0.55000kg
Description
Absorb adequate amounts of buffer for semi-dry electroblotting without the loss of mechanical stability with Thermo Scientific™ Owl™ Blotting Paper, high-quality blotting paper ideal for wicking and blotting applications.
- High purity, 100% cotton fiber
- Available in pre-cut mini gel and large format gel sizes
- Compatible with Owl electroblotting systems
Ordering Information:
- Sold in packs of 100 sheets
Specifications
10 cm | |
Sheets | |
Owl electroblotting systems | |
Sheet | |
10 cm x 10 cm | |
Owl |
10 cm | |
35 mL/min. | |
183 g/m² | |
100% Cotton Fiber | |
0.83 mm |