Learn More
Kartell™ Overturnable Stoppers
Product Code. 10059632
Change view
Click to view available options
Material:
Rubber
Unit Size:
Pack of 10
Product Code. Material unitSize
10059632 Rubber Pack of 10
1 options
Product Code. 10059632 Supplier Kartell™ Supplier No. 0119500
Only null left
Add to basket
Add to basket
Pierceable stoppers to withdraw solutions and suspensions from closed containers, using syringes
  • Sylicon rubber
  • Suitable for use in food and beverage industry
  • Specifications
    Height (Metric) Overturnable Section: 18 mm Lower Section: 12 mm
    Diameter (Metric) Bottom 20 mm
    Material Rubber