missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Organic Acids Cuvette Test

Product Code. 10172344
Change view
Click to view available options
No. of Tests:
25
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Tests unitSize
10172344 25 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10172344 Supplier Hach Lange™ Supplier No. LCK365

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Order Info

dimRange

TRUSTED_SUSTAINABILITY

Specifications

No. of Tests 25
Range 50-2500mg/L
Storage Requirements 15°C to 25°C
Test Method Esterification
Type Organic Acids Cuvette Test
compliance-icons compliance-icons
Product Identifier
  • Organische Säuren/Organic Acids/Acides organiques
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: ethelene glycol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.