Learn More
Hamilton™ One-Hub Assembled Tubing Assemblies
Product Code. 10106833
Change view
Click to view available options
Needle Gauge:
8 G
10 G
11 G
12 G
13 G
14 G
16 G
17 G
19 G
20 G
26 G
30 G
Length (Metric):
304.8 mm
Custom
Unit Size:
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Length (Metric) unitSize
10106833 10 G 304.8 mm Each
10394703 11 G 304.8 mm Each
11543084 12 G 304.8 mm Each
10170883 13 G 304.8 mm Each
11583124 20 G Custom Each
11573124 20 G Custom Each
11563084 14 G 304.8 mm Each
11563124 19 G Custom Each
11593134 12 G Custom Each
11593084 17 G 304.8 mm Each
11503134 26 G Custom Each
11523134 30 G Custom Each
11503084 8 G 304.8 mm Each
11533144 16 G Custom Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 10106833 Supplier Hamilton™ Supplier No. 90610
Providing a variety of connection solutions in the laboratory.
  • Fitting manufactured from CTFE
  • Tubing manufactured from PTFE
  • For female luers
  • A variety of gauges and lengths available
Specifications
Type Tubing
Material PTFE
Length (Metric) 304.8 mm
Needle Gauge 10 G