missing translation for 'onlineSavingsMsg'
Learn More

Omethoate-d6 (O,O-dimethyl-d6), CDN

Product Code. 30320843
Change view
Click to view available options
Quantity:
0.01 g
Unit Size:
0.01g
Product Code. Quantity unitSize
30320843 0.01 g 0.01g
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30320843

Brand: CDN CDND72640.01G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material OmethoateD6(dimethylD6)
CAS 1219804-92-8
Molecular Formula C5 2H6 H6 N O4 P S
Quantity 0.01 g
Synonym Omethoate D6 (O-dimethyl D6), 2-[Bis(trideuteriomethoxy)phosphorylsulfanyl]-N-methyl-acetamide, Omethoate D6 (dimethyl D6)
InChI Formula InChI=1 S/C5H12NO4PS/c1-6-5(7)4-12-11(8,9-2)10-3/h4H2,1-3H3,(H,6,7)/i2D3,3D3
SMILES [2 H]C([2 H])([2 H])OP(=O)(OC([2 H])([2 H])[2 H])SCC(=O)NC
IUPAC Name 2-[bis(trideuteriomethoxy)phosphorylsulfanyl]-N-methylacetamide
Molecular Weight (g/mol) 219.23
Formula Weight 219.0601 g/mol
Percent Purity 99 atom % D, min 95% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.