Learn More
Bohlender™ O-ring
Specifications
Specifications
| For Use With (Application) | Used as sealing |
| Quantity | 1Each |
| Product Type | O-Ring |
| Material | Silicone core/FEP Coating |
| Size | 132 x 4mm |