missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ O-1602

Description
Analog of cannabidiol that is a potent agonist at the GPR55 cannabinoid receptor (EC50 values are 13, > 30000 and > 30000 nM for GPR55, CB1 and CB2 receptors respectively). Induces activation of RhoA, cdc42 and rac1. Increases proliferation of neural stem cells NSCs in vitro and in vivo. Also promotes neurogenesis in vivo.
Specifications
Specifications
| CAS | 317321-41-8 |
| Quantity | 10 mg |
| Molecular Formula | C17H22O2 |
| Formula Weight | 258.36 |
| Purity | >98% |
| Molecular Weight (g/mol) | 258.361 |
| Chemical Name or Material | O-1602 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?