missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Plasmid DNA

Product Code. 11912252
Change view
Click to view available options
No. of Reactions:
10 Preps
250 Preps
50 Preps
Includes:
Lid
No Lid
Unit Size:
10 reactions
250 reactions
50 reactions
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Includes unitSize
11912252 250 Preps No Lid 250 reactions
11942502 10 Preps Lid 10 reactions
11932392 50 Preps Lid 50 reactions
12353358 250 Preps Lid 250 reactions
11942362 50 Preps No Lid 50 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11912252 Supplier Macherey-Nagel™ Supplier No. 740499.250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Prepare small scale high-purity plasmid DNA rapidly. NucleoSpin™ Plasmid Columns come with or without a cap lid.

Features:

  • For rapid small scale preparation high-purity plasmid DNA
  • Fast procedure – 6 Minipreps in 25 min.
  • High capacity – up to 50μg of plasmid DNA
  • Chaotropic salt wash buffer – extra pure DNA

Applications:

  • High-copy plasmids from E. coli
  • Low-copy plasmids, P1 constructs, or cosmids (increased buffer volumes required)
  • Plasmid DNA from Gram-positive bacteria and plasmid DNA clean-up from reaction mixtures
  • Supplementary protocol for M13 DNA available on request
  • Typical downstream applications: cloning, sequencing, PCR, transformation, restriction analysis

Specifications

Format Column
Isolation Technology Silica Membrane Technology
Sequencing Type DNA and RNA Sequencing
For Use With (Application) High-copy plasmids from E. coli
Includes No Lid
No. of Reactions 250 Preps
Product Type Plasmid Columns
Quantity 2 mL
Sample Volume 2 to 10 mL
Size < 50 kbp
Test Time 25min.
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide solution (diluted < 2 %); guanidine hydrochloride; 2-propanol;Rnase
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.