missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Plasmid Columns for plasmid DNA
Description
NucleoSpin™ Plasmid
- For rapid small scale preparation high-purity plasmid DNA
- Fast procedure – 6 Mini preps in 25 min
- High capacity – up to 50 μg of plasmid DNA
- Chaotropic salt wash buffer – extra pure DNA
- Downstream applications: cloning, sequencing, PCR, transformation, restriction analysis
- High-copy plasmids from E. coli
- Low-copy plasmids, P1 constructs, or cosmids (increased buffer volumes required)
- Plasmid DNA from Gram-positive bacteria and plasmid DNA clean-up from reaction mixtures
- Supplementary protocol for M13 DNA availabl
Specifications
Specifications
| Format | Mini Spin Column |
| Isolation Technology | Silica Membrane Technology |
| Sample Type | E. coli Culture |
| Efficiency | Binding capacity: 50 μg |
| Elution Volume | 50 μL |
| Final Product Type | Sequencing-grade plasmid DNA |
| For Use With (Equipment) | NucleoSpin™ Plasmid Kit |
| For Use With (Application) | Preparation of high-purity plasmid DNA |
| Includes | Columns only |
| Product Type | NucleoSpin Plasmid lidded Column |
| Show More |
Safety and Handling
Product Identifier
- NucleoSpin Plasmid
- MIXTURE LIST-Contains : sodium hydroxide solution (diluted < 2 %), dodecyl sulfate,sodium salt/guanidine hydrochloride/2-pr
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?