missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ RNA/DNA 400, Maxi kit with anion exchange technology

Product Code. 10181291
Change view
Click to view available options
:
No. of Reactions: 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10181291 No. of Reactions: 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10181291 Supplier Macherey-Nagel™ Supplier No. 740651

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Anion exchange chromatography for nucleic acids of highest integrity.

NucleoBond™ RNA/DNA 80 and 400 kits are designed for the isolation and purification of up to 80μg or 400μg ultra pure total RNA. A special manufacturing process guarantees RNase-free handling. Due to the superior resolving power of NucleoBond™ anion exchanger contaminations are washed away, and RNA and genomic DNA can be eluted separately.

Specific references:

M. Jiang et al. , J. Virol. Methods

38

(1992) 205T. Roitsch et al. , Plant Physiol.

108

(1995) 285D. Godt et al. , J. Plant Physiol.

146

(1995) 231R. Ehness et al. , The Plant Cell

9

(1997) 1825S. P. Hale et al. , Science

276

(1997) 1250Kit components: NucleoBond™ RNA/DNA 80 - NucleoBond™ AXR 80 columns, all required buffers. NucleoBond™ RNA/DNA 400 - NucleoBond™ AXR 400 columns, all required buffers.

  • Anion-exchange technology
  • Typical RNA yield: 70μg to 300μg from cultured cells 30μg to 150μg from tissue 50μg to 200μg from bacteria
  • Sample material: eukaryotic cultured cells, tissue, bacteria/yeast cells
  • Fragment size: 50bp to 300kb
  • Binding capacity: 80μg for NucleoBond RNA/DNA 80 400μg for NucleoBond RNA/DNA 400
  • Preparation time: 1.5 hr. to 2. 5 hr.
  • Format: gravity-flow columns - midi or maxi

Specifications

No. of Reactions 10 Preps
Product Type RNA/DNA 400
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, guanidiniumthiocyanate, tritonx-100
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.