missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoBond™ CB 500, Midi kit for high integrity DNA from cells and blood
Description
Genomic DNA quality is extraordinarily high. Samples are first lysed under special conditions for removal of erythrocytes. After incubation with proteinase K, the sample is diluted with buffer N2 before loading onto the NucleoBond™ column. After elution with high-salt buffer the genomic DNA is precipitated with isopropanol. The DNA purified by this method is suitable for all kinds of subsequent reactions.Kit components: NucleoBond™ CB 100 kit - NucleoBond AXG 100 columns, buffers, proteinase K. NucleoBond™ CB 500 kit - NucleoBond™ AXG 500 columns, buffers, proteinase K.
- Anion exchange technology
- Format: gravity flow columns
- Sample material: whole blood, buffy coat, cultured cells (up to 20 mL blood)
- Fragment size: 500bp to 300kb
- Yield: 100μg or 500μg depending on column used
- Binding capacity: 100μg or 500μg depending on column used
- Preparation time: 4 to 5hrs
Specifications
Specifications
| Format | Midi Gravity-flow Column |
| Isolation Technology | Anion Exchange Chromatography Technology |
| For Use With (Application) | For High-copy Plasmids From E. coli |
| No. of Reactions | 10 |
| Product Type | NucleoBond™ AXG 500 Columns |
| Test Time | 4 to 5hr. |
| Yield | 500μg |
Safety and Handling
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?