missing translation for 'onlineSavingsMsg'
Learn More

Showa Best Gloves™ NSK26™ Coated Nitrile Gloves

Product Code. 12980775
Click to view available options
Size:
8
10
11
Unit Size:
Pack of 72
This item is not returnable. View return policy

Product Code. 12980775

Brand: Showa Best Gloves™ NSK2608S

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

SHOWA™ NSK26™ coated nitrile gloves protect the hand and arm from a wide range of chemical splashes whilst providing comfort and touch sensitivity

  • Fully coated nitrile glove that absorbs perspiration for ultra-comfortable extended wear and with good mechanical resistance
  • Cut and sewn construction designed for easy movement and continuous wear
  • Protects the hand from oils, hydrocarbons, grease and abrasion, with effective long lasting textured grip
  • Extended protection on the forearm
  • Impermeable for working in damp or greasy environments
  • Surface provides tactile feel and better grip
  • Lower allergy risks
  • Suitable for use in agriculture, construction, food handling, forestry & fishing, manufacturing, municipal services, oil and gas, wet work

Specifications

Certifications/Compliance EN 374, EN388, FOOD CONTACT DIRECTIVE 2002 / 72 EC
Color Blue
Exterior Finish Rough
Material Nitrile
Powdered Powder Free
Size 8
Sterility Non-sterile
For Use With (Application) Agrochemicals, chemical handling, paints, oils, industrial tool maintenance, refining, waste and water treatment
Glove Cuff Style Elasticated
Liner Material Cotton/Polyester
Length (Metric) 650 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.