missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NPR3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10181410
Change view
Click to view available options
Quantity:
200 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10181410 200 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10181410 Supplier Invitrogen™ Supplier No. PA515394

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NPR3 Polyclonal Antibody for ICC/IF, IHC (P), Western Blot

This gene encodes one of three natriuretic peptide receptors. Natriutetic peptides are small peptides which regulate blood volume and pressure, pulmonary hypertension, and cardiac function as well as some metabolic and growth processes. The product of this gene encodes a natriuretic peptide receptor responsible for clearing circulating and extracellular natriuretic peptides through endocytosis of the receptor. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPR3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene NPR3
Gene Accession No. P17342
Gene Alias A NPR-C; ANP-C; ANP-CR; ANPRC; ANPR-C; atrial natriuretic peptide clearance receptor; Atrial natriuretic peptide clearence receptor 3; atrial natriuretic peptide C-type receptor; atrial natriuretic peptide receptor 3; atrial natriuretic peptide receptor 3-like; atrial natriuretic peptide receptor type C; atrionatriuretic peptide receptor C; C5orf23; EF-2; guanylate cyclase C; GUCY2B; lgj; longjohn; natriuretic peptide receptor 3; natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptide receptor C); Nppc receptor; Npr3; NPRC; NPR-C; stri; strigosus
Gene Symbols NPR3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 67-97 amino acids from the N-terminal region of human Natriuretic Peptide Receptor C.
Purification Method Ammonium sulfate precipitation
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4883
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.