Learn More
Invitrogen™ TMB Solution (BLOT)
TMB (Tetramethyl Benzidine) chromogen solution
Brand: Invitrogen™ 002019
Description
May be used with horseradish peroxidase (HRP)-based immunodetection procedures
Proteins, Expression, Isolation and Analysis, Western Blotting
Order Info
Shipping condition: Wet ice
Specifications
| Store at 2-8°C | |
| Colorimetric | |
| Wet Ice |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
For Research Use Only. Not for use in diagnostic procedures.