missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Norepinephrine bitartrate monohydrate, MedChemExpress
Description
Norepinephrine (Levarterenol
Specifications
Specifications
| Chemical Name or Material | Norepinephrine bitartrate monohydrate |
| CAS | 108341-18-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C12H19NO10 |
| Quantity | 500 mg |
| Synonym | Levarterenol bitartrate monohydrateL-Noradrenaline bitartrate monohydrate |
| Solubility Information | H2O : 50 mg/mL (148.24 mM; Need ultrasonic) ∣DMSO : ≥ 30 mg/mL (88.95 mM) |
| SMILES | OC1=CC=C([C@@H](O)CN)C=C1O.O=C(O)[C@H](O)[C@@H](O)C(O)=O.O |
| Molecular Weight (g/mol) | 337.28 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?