missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente™ Nitrogen Round-Bottom Flasks with NS Stopcock

Product Code. 10070231
Change view
Click to view available options
:
Capacity (Metric): 250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10070231 Capacity (Metric): 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10070231 Supplier Lenz Laborglasintrumente™ Supplier No. 03360449
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Nitrogen round-bottom flask (to Schlenk) with NS stopcock

  • With conical ground joint and NS stopcock
  • With inscription label
  • Material: DURAN™

Specifications

Description Stopcock bore - 2.5 mm
Capacity (Metric) 250 mL
Material Borosilicate Glass (3.3)
No. per Pack 1
Socket Joint NS 29/32
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.