missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) Sulfate Heptahydrate, purum p.a; ≥99.0% (KT), Honeywell™

Product Code. p-7120622 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15666890 250 g Poly bottle 250g
15656890 1 kg Poly bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15666890 Supplier Honeywell Research Chemicals Supplier No. 72285250G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a., crystallized,≥99.0%

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-98-1
Molecular Formula H14NiO11S
Molecular Weight (g/mol) 280.85
MDL Number MFCD00149814
InChI Key OGKAGKFVPCOHQW-UHFFFAOYSA-L
Synonym nickel ii sulfate heptahydrate, nickel sulfate heptahydrate, nickel 2+ sulfate heptahydrate, nickelous sulfate heptahydrate, unii-596idd57nr, sulfuric acid, nickel 2+ salt 1:1 , heptahydrate, nickel sulfate heptahydrate nickel and nickel compounds, ni.so4.7h2o, ksc492q3n, nickel sulfate-water 1/7
PubChem CID 61474
ChEBI CHEBI:53504
IUPAC Name nickel(2+);sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O

Specifications

Physical Form Crystalline
Quantity 250 g
Linear Formula NiSO4 · 7H2O
UN Number UN3077
Formula Weight 280.86g/mol
Percent Purity ≥99.0% (KT)
Packaging Poly bottle
Density 1.948g/mL at 25°C
Chemical Name or Material Nickel(II) sulfate heptahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.