Learn More
Invitrogen™ NGFR Monoclonal Antibody (MC192)
Description
Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. The MC192 is supplied in lyophilised form from Protein G-purified hybridoma cell culture supernatants. The lyophilised antibody is stable when stored at 2-8 °C or -20 °C. After reconstitution undiluted aliquots should be kept at -20 °C for up to six months. For additional stability Glycerol (1:1) may be added after reconstitution. Repetitive freeze/thaw cycle should be avoided.
Specifications
Specifications
| Antigen | NGFR |
| Applications | ELISA, Immunohistochemistry (Frozen), Western Blot |
| Classification | Monoclonal |
| Clone | MC192 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | Ngfr |
| Gene Accession No. | P07174 |
| Gene Alias | CD271; CD271 antigen; Gp80-LNGFR; LNGFR; low affinity nerve growth factor receptor; low affinity neurotrophin receptor p75NTR; LOW QUALITY PROTEIN: tumor necrosis factor receptor superfamily member 16; low-affinity nerve growth factor receptor; nerve growth factor receptor; nerve growth factor receptor (TNFR superfamily, member 16); nerve growth factor receptor, fast; NGF receptor; NGFR; NGR; p75; p-75; p75 ICD; p75 neurotrophin receptor; p75 ntr; p75(NTR); p75NGFR; p75NTR; RNNGFRR; TNFR superfamily, member 16; Tnfrsf16; TNR16; Tumor necrosis factor receptor superfamily member 16 |
| Gene Symbols | Ngfr |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.