missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dr.Weigert™ Neodisher Z
Description
- Based on citric acid
- neodisher Z is used for neutralizing alkaline residues in washer disinfection
- Without phosphates, phosphoric acid, nitrogens compounds and surfactants
- Dosage of between 2-4 mL/L
- Gentle on materials
- Laboratory glassware in medical, industrial and water laboratories
Specifications
Specifications
| Product Type | Neodisher Z |
| Form | Liquid |
| pH | 3 to 2.6 |
| Sterility | Non-sterile |
| Composition | Citric acid, 2-octyl-2 H-isothiazol-3-one |
| Concentration | 1 to 5 mL/L |
| Container Type | Canister (Elongated) |
| Density | 1.2 g/cm3 (20°C) |
| Description | 220 x 240 x 140 mm |
| Filtered | Filtered |
| Show More |
Safety and Handling
Product Identifier
- neodisher Z
- Warning
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin sensitiser Category 1
- H319-Causes serious eye irritation.
- H317-May cause an allergic skin reaction.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: citric acid, anhydrous
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?