missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NEFM Monoclonal Antibody (RMO 14.9)
GREENER_CHOICE

Product Code. 10017952
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10017952 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10017952 Supplier Invitrogen™ Supplier No. 341000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

NEFM Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Neurofilaments (NFs) are a type of intermediate filament (IF) expressed almost exclusively in neuronal cells, and in those cells most prominently in large axons. NFs, in most vertebrates, are composed of three different polypeptide chains with different molecular weights - neurofilament medium protein (NF-M), high (NF-H) and light protein (NF-L), which share sequence and structural similarity in a coiled-coil core domain, but differ in the length and sequence of their N-termini and more dramatically of their C-termini which in the case of NF-M and NF-H form the flexible extensions that link NFs to each other and to other elements in the cytoplasm. NF-M protein tail-mediated interactions of neurofilaments are critical for size and cytoskeletal architecture of axons, and are mediated, in part, by the highly phosphorylated tail domain of this protein. NF-M phosphorylation and O-GlcNAcylation are regulated reciprocally and affect its translocation and filament formation and function. Antibodies to the various neurofilament subunits are very useful cell type markers since the proteins are among the most abundant of the nervous system, are expressed only in neurons and are biochemically very stable.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEFM
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone RMO 14.9
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene NEFM
Gene Accession No. P07197, P08553, P12839, P16053, P54938
Gene Alias 160 kDa neurofilament protein; major neuronal intermediate filament subunit; medium neurofilament subunit; middle molecular weight neurofilament protein NF-M(2); Nef3; Nefm; nef-m; nefm.S; nefm-a; nefm-b; Neurofilament 3; neurofilament 3 (150kDa medium); neurofilament 3, medium; neurofilament M subunit; neurofilament medium; neurofilament medium polypeptide; neurofilament medium S homeolog; neurofilament medium tail domain; neurofilament protein M; Neurofilament protein, middle polypeptide; neurofilament triplet M protein; neurofilament, medium polypeptide; neurofilament, medium polypeptide 150kDa; neurofilament, medium polypeptide S homeolog; neurofilament-3 (150 kD medium); neurofilament-M; neurofilament-M subunit; neuronal intermediate filament; NF160; NF165; NFM; NF-M; NF-M c-terminus; NF-M protein; NMC; XELAEV_18020229mg
Gene Symbols NEFM
Host Species Mouse
Immunogen Rat NF-M.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009343, 18040, 24588, 396206, 4741
Target Species Human, Mouse, Rat, Rabbit, Chicken
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.