missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ PPCO Conical-Bottom Centrifuge Tubes

Description
Simplify measurements using Thermo Scientific™ Nalgene™ PPCO Conical-Bottom Centrifuge Tubes, which feature molded-in graduations. Translucent polypropylene copolymer (PPCO) has excellent chemical resistance.
Features:
- Translucent Polypropylene Copolymer (PPCO) is compatible with a wide range of lab reagents
- Designed for low-speed centrifugation up to 6000 × g in refrigerated or non-refrigerated centrifuges (tubes must be filled at least 80% for proper performance)
- Conical bottoms concentrate pellet in a small area for easy isolation and retrieval
- Molded-in graduations last the life of the tube
- Last longer than polycarbonate tubes under conditions of repeated autoclaving
- Tube are both autoclavable and graduated
Ordering Information:
Supplied without closure; friction fit closures available. For 15mL use DS3111-00017; for 50mL use DS3111-0029.
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Material | Polypropylene Copolymer |
| No. per Case | 100 |
| No. per Pack | 10 |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Shape | Conical |
| Length (Metric) | 133.1 mm |
| Bottom Shape | Conical |
| Writing Area | No |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?