missing translation for 'onlineSavingsMsg'
Learn More

N6-(4-Hydroxybenzyl)adenosine, MedChemExpress

Product Code. 16497628
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497628 10 mg 10mg
16487536 100 mg 100mg
16413647 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16497628 Supplier Medchem Express Supplier No. HY18775

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress N6-(4-Hydroxybenzyl)adenosine is a inhibitor of platelet aggregation induced in vitro by collagen and their activity range was demonstrated (IC50: 6.77-141 μM).

N6-(4-Hydroxybenzyl)adenosine is a inhibitor of platelet aggregation induced in vitro by collagen and their activity range was demonstrated (IC50: 6.77-141 μM).

Specifications

Chemical Name or Material N6-(4-Hydroxybenzyl)adenosine
CAS 110505-75-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H19N5O5
Quantity 10 mg
Synonym Para-topolin riboside
Solubility Information DMSO : ≥ 100 mg/mL (267.84 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=CN=C3NCC4=CC=C(O)C=C4)O1)O)O
Molecular Weight (g/mol) 373.36
Formula Weight 373.36
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.