missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butyl-alpha-phenylnitrone, 98%

Product Code. 10046333 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10046333 5 g Glass bottle 5g
10783561 1 g Glass bottle 1g
11964571 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10046333 Supplier Thermo Scientific Acros Supplier No. 177350050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3376-24-7 | C11H15NO | 177.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3376-24-7
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.25
MDL Number MFCD00008799
InChI Key IYSYLWYGCWTJSG-FMIVXFBMSA-N
Synonym e-n-benzylidene-2-methylpropan-2-amine oxide
PubChem CID 10313352
IUPAC Name N-tert-butyl-1-phenylmethanimine oxide
SMILES CC(C)(C)[N+](=CC1=CC=CC=C1)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 71.0°C to 75.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH=N(O)C(CH3)3
Quantity 5 g
Merck Index 15,7162
Formula Weight 177.25
Percent Purity 98%
Chemical Name or Material N-tert-Butyl-α-phenylnitrone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.