missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N,N-Dimethylglycine hydrochloride, 99%

Product Code. 10026293 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10026293 100 g Glass bottle 100g
10431441 5 g Glass bottle 5g
10378290 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10026293 Supplier Thermo Scientific Chemicals Supplier No. 204661000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2491-06-7 | C4H9NO2·HCl | 139.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2491-06-7
Molecular Formula C4H9NO2·HCl
Molecular Weight (g/mol) 139.58
MDL Number MFCD00012610
InChI Key FKASAVXZZLJTNX-UHFFFAOYSA-N
Synonym n,n-dimethylglycine hydrochloride, dimethylglycine hydrochloride, glycine, n,n-dimethyl-, hydrochloride, n-methylsarcosine hydrochloride, n,n-dimethylglycine hcl, dimethylamino acetic acid hydrochloride, unii-yxk75eae92, yxk75eae92, n,n-dimethylaminoacetic acid hydrochloride, 2-dimethylamino acetic acid hydrochloride
PubChem CID 75605
IUPAC Name 2-(dimethylamino)acetic acid;hydrochloride
SMILES CN(C)CC(=O)O.Cl

Specifications

Melting Point 188°C to 191°C
Color White
CAS Min % 98.5
CAS Max % 100.0
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99%
Linear Formula (CH3)2NCH2CO2H·HCl
Beilstein 04, I, 469
Merck Index 15, 3269
Solubility Information Solubility in water: 100g/L (20 c). Other solubilities: insoluble in chloroform and acetone
Formula Weight 139.58
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material N, N-Dimethylglycine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.