missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylformamide, 99%

Product Code. 10174100 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10174100 100 mL Glass bottle 100mL
10713924 25 mL Glass bottle 25mL
10657364 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10174100 Supplier Thermo Scientific Acros Supplier No. 181371000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 617-84-5 | C5H11NO | 101.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-84-5
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00003287
InChI Key SUAKHGWARZSWIH-UHFFFAOYSA-N
Synonym diethylformamide, formamide, n,n-diethyl, n-formyldiethylamine, diethyl formamide, formyldiethylamine, diethylamid kyseliny mravenci, n,n-diethylcarboxamide, diethylamid kyseliny mravenci czech, diethyl-formamide, formamide, diethyl
PubChem CID 12051
IUPAC Name N,N-diethylformamide
SMILES CCN(CC)C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9000g/mL
Boiling Point 176.0°C to 177.0°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HCON(C2H5)2
Refractive Index 1.4330 to 1.4350
Quantity 100 mL
Beilstein 04, 109
Specific Gravity 0.9
Formula Weight 101.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Diethylformamide, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N-Diethylformamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: N,N-Diethylformamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.