missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Diacetyl-L-cystine Dimethyl Ester, Mikromol™
Description
N,N'-Diacetyl-L-cystine Dimethyl Ester
Specifications
Specifications
| Chemical Name or Material | N,N'-Diacetyl-L-cystine Dimethyl Ester |
| CAS | 32381-28-5 |
| Analyte or Component Names | N,N'-Diacetyl-L-cystine Dimethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H20 N2 O6 S2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | N,N'-Diacetyl-L-cystine 1,1'-dimethyl ester, Cystine, N,N'-diacetyl-, dimethyl ester, L- (6CI,7CI,8CI), L-Cystine, N,N'-diacetyl-, dimethyl ester (9CI), L-N,N'-Diacetylcystine dimethyl ester, N,N'-Diacetylcystine dimethyl ester, NSC 136019 |
| InChI Formula | InChI=1S/C12H20N2O6S2/c1-7(15)13-9(11(17)19-3)5-21-22-6-10(12(18)20-4)14-8(2)16/h9-10H,5-6H2,1-4H3,(H,13,15)(H,14,16)/t9-,10-/m0/s1 |
| SMILES | COC(=O)[C@H](CSSC[C@H](NC(=O)C)C(=O)OC)NC(=O)C |
| IUPAC Name | methyl (2R)-2-acetamido-3-[[(2R)-2-acetamido-3-methoxy-3-oxopropyl]disulfanyl]propanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?