missing translation for 'onlineSavingsMsg'
Learn More

N-(alpha-Phenylethyl)ibuprofenamide, Mikromol™

Product Code. 17990796
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity unitSize
17990796 25 mg 25mg
1 options
This item is not returnable. View return policy

Product Code. 17990796

Brand: Mikromol MM0002.410025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-(alpha-Phenylethyl)ibuprofenamide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(1-Phenylethyl) Ibuprofen Amide(mixture of diastereomers)
CAS 105959-56-6
Analyte or Component Names N-(1-Phenylethyl) Ibuprofen Amide(mixture of diastereomers)
Shipping Condition Room Temperature
Molecular Formula C21 H27 N O
Synonym Benzeneacetamide, α-methyl-4-(2-methylpropyl)-N-(1-phenylethyl)-, 2-(4-isobutylphenyl)-N-(1-phenylethyl)propanamide, N-(α-Phenylethyl)ibuprofenamide
InChI Formula InChI=1S/C21H27NO/c1-15(2)14-18-10-12-19(13-11-18)16(3)21(23)22-17(4)20-8-6-5-7-9-20/h5-13,15-17H,14H2,1-4H3,(H,22,23)
SMILES CC(C)Cc1ccc(cc1)C(C)C(=O)NC(C)c2ccccc2
IUPAC Name 2-[4-(2-methylpropyl)phenyl]-N-(1-phenylethyl)propanamide
Molecular Weight (g/mol) 309.45
Quantity 25 mg
Formula Weight 309.2093 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.