missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Acetylethanolamine, 96%

Artikelnummer. 10173291 Shop alle Thermo Scientific Chemicals produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
5 g
100 g
Packaging:
Glass bottle
Pakningsstørrelse:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packaging unitSize
10173291 5 g Glass bottle 5g
10411202 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 10173291 Leverandør Thermo Scientific Acros Leverandørnr. 146200050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

CAS: 142-26-7 | C4H9NO2 | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 142-26-7
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00002836
InChI Key PVCJKHHOXFKFRP-UHFFFAOYSA-N
Synonym n-2-hydroxyethyl acetamide, n-acetylethanolamine, 2-acetamidoethanol, acetamide, n-2-hydroxyethyl, acetylcolamine, 2-acetylaminoethanol, acetamide mea, n-ethanolacetamide, n-acetyl-2-aminoethanol, hydroxyethyl acetamide
PubChem CID 8880
ChEBI CHEBI:74687
IUPAC Name N-(2-hydroxyethyl)acetamide
SMILES CC(=O)NCCO

Tekniske data

CAS Min % 95.0
CAS Max % 100.0
Melting Point 15.8°C
Color White to Yellow
Density 1.1100g/mL
Boiling Point 135°C to 137°C (3.0 mmHg)
Flash Point 176°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3CONHCH2CH2OH
Refractive Index 1.4720 to 1.474
Quantity 5 g
Beilstein 04, I, 430
Specific Gravity 1.11
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in alcohol and ether
Formula Weight 103.12
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material N-Acetylethanolamine
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • N-Acetylethanolamine
Signalord
  • Warning
Farekategori
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sikkerhedssætning
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.