missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MTHFR Recombinant Rabbit Monoclonal Antibody (K01_1U19)
Description
MTHFR Recombinant Monoclonal Antibody for ICC/IF, Western Blot
The protein encoded by this gene catalyzes the conversion of 5,10-methylenetetrahydrofolate to 5-methyltetrahydrofolate, a co-substrate for homocysteine remethylation to methionine. Genetic variation in this gene influences susceptibility to occlusive vascular disease, neural tube defects, colon cancer and acute leukemia, and mutations in this gene are associated with methylenetetrahydrofolate reductase deficiency.
Specifications
Specifications
| Antigen | MTHFR |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | K01_1U19 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4 |
| Gene | MTHFR |
| Gene Accession No. | P42898, Q9WU20 |
| Gene Alias | 5,10-methylenetetrahydrofolate reductase; 5,10-methylenetetrahydrofolate reductase (NADPH); AI323986; Methylenetetrahydrofolate reductase; methylenetetrahydrofolate reductase (NAD(P)H); MTHFR; RP11-56N19.4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?