missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat Set

Product Code. 10053392
Change view
Click to view available options
Target:
Human Set
Human Single
Mouse Set
Mouse Single
Rat Single
Quantity:
1 Set
20 nmol
20 x 3 nmol
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Target Quantity unitSize
10053392 Mouse Single 20 nmol Each
10347922 Rat Single 20 nmol Each
10337922 Mouse Set 20 x 3 nmol Each
10083282 Human Single 20 nmol Each
10093282 Human Set 1 Set Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10053392 Supplier Invitrogen™ Supplier No. 1320001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Stealth RNAi Predesigned siRNAs are designed to provide robust and long-lasting gene silencing with minimal off-target effects. Stealth RNAi siRNAs are modified synthetic 25-mer RNA duplexes. Our proprietary technology ensures high specificity and stability, making Stealth RNAi siRNAs an ideal choice for researchers seeking reliable and efficient gene knockdown in human, mouse, or rat.

Stealth RNAi Predesigned siRNAs enable robust and long-lasting gene silencing with reduced off-target effects. Stealth RNAi siRNAs are modified synthetic 25-mer RNA duplexes. Our proprietary technology helps ensure high specificity and stability, making Stealth RNAi siRNAs an excellent choice for researchers seeking reliable and efficient gene knockdown in human, mouse, or rat.

Key features include:

Higher specificity and potency—algorithm designs minimize off-target effects and maximize target gene knockdown

Enhanced stability—engineered for enhanced stability in serum and cellular environments, ensuring prolonged gene silencing

Minimal cellular toxicity—designed to evade immune detection, for reduced risk of immune response

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Description Stealth RNAi™ Pre-designed siRNA Tube, 20nmol, Mouse Single Target, Good Knockdown, Low Off-target Effects, Standard Purity, Used For Treating Parkinson's Disease
Format Tube
RNAi Type siRNA, miRNA
Final Product Type Stealth RNAi™ Pre-designed siRNA Tube
For Use With (Application) For treating Parkinson's disease
Product Type Stealth RNAi™ Pre-designed siRNA Tube
Purity Standard purity
Quantity 20 nmol
Sample Size 20nmol
Target Mouse Single
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.