missing translation for 'onlineSavingsMsg'
Learn More

Mouse Mast Cell Protease-1/Mcpt1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. p-7066343
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18713529 25 μg 25µg
18003452 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18713529 Supplier R&D Systems Supplier No. MAB5146SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Mast Cell Protease-1/Mcpt1 Monoclonal specifically detects Mast Cell Protease-1/Mcpt1 in Mouse samples. It is validated for Western Blot, Simple Western.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mast Cell Protease-1/Mcpt1
Applications Western Blot
Classification Monoclonal
Clone 285008
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P11034
Gene Alias AV080368, Mast Cell Protease1, Mcp1, Mcp-1, MMPC-1
Gene Symbols Mcpt1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Mast Cell Protease-1/Mcpt1 Ile21-Lys246 Accession # P11034
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17224
Test Specificity Detects mouse Mast Cell Protease-1/Mcpt1 in direct ELISAs and Western blots. Detects rat Mast Cell Protease-1/Mcpt1 in Simple Western. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) MCP-6, -7, or -11 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.