Learn More
Gibco™ Mouse CXCL12 (SDF-1b) Recombinant Protein
Description
Carrier-Free Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute in sterile, distilled water to a concentration of 0.1-1.0 mg/mL. Further dilutions should be made in aqueous buffers supplemented with carrier protein, such as 0.1-1.0% BSA. Storage: This lyophilized preparation is stable at 2°C to 8°C, but should be kept at -20°C for long term storage, preferably desiccated. Upon reconstitution, the preparation is stable for up to one week at 2°C to 8°C. For maximal stability, apportion the reconstituted preparation into working aliquots and store at -20°C. Avoid repeated freeze/thaw cycles.
Specifications
Specifications
| Accession Number | P40224 |
| For Use With (Application) | Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 20315 |
| Molecular Weight (g/mol) | 8.5 kDa |
| Name | Mouse CXCL12 (SDF-1b) |
| Purification Method | Purified |
| Quantity | 10 μg |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.