missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse CXCL12 (SDF-1a) Recombinant Protein

Product Code. 10100593
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10100593 10 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10100593 Supplier Gibco™ Supplier No. PMC1354

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

Reconstitution: We recommend that the vial be briefly centrifuged prior to opening to bring the contents to the bottom. This lyophilized protein should be reconstituted in sterile distilled water to a concentration of 0.1-1.0 mg/mL. Further dilutions should be made in tissue culture grade buffered solutions with heat inactivated fetal calf serum or low endotoxin BSA. Storage: Lyophilized human SDF-1 alpha should be stored at 2°C to 8°C, preferably desiccated. Store reconstituted human SDF-1 alpha at < -20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.

CXCL12 is a stromal cell-derived alpha chemokine member of the intercrine family. This gene product and its receptor CXCR4 can activate lymphocytes and have been implicated in the metastasis of some cancers such as breast cancer. Mutations in this gene are associated with resistance to human immunodeficiency virus type 1 infections. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P40224
For Use With (Application) Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 20315
Molecular Weight (g/mol) 7.9 kDa
Name Mouse CXCL12 (SDF-1a)
Purification Method Purified
Quantity 10 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <0.1 ng/μg
Gene Alias 12-O-tetradecanoylphorbol 13-acetate repressed protein 1; AI174028; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 12; chemokine (C-X-C motif) ligand 12 (stromal cell-derived factor 1); CXC; CXC motif chemokine; C-X-C motif chemokine 12; C-X-C motif chemokine ligand 12; CXCL; Cxcl12; hIRH; hSDF-1; Intercrine reduced in hepatomas; IRH; PBSF; pre-B cell growth-stimulating factor; pre-B-cell growth-stimulating factor; Scyb12; Sdf1; SDF-1; SDF-1 gamma; SDF1A; SDF-1a; SDF-1-alpha(3-67); SDF1B; SDF-1b; SDF-1-beta(3-72); Stromal cell-derived factor 1; stromal cell-derived factor-1 alpha; stromal cell-derived factor-1 gamma; thymic lymphoma cell-stimulating factor; TLSF; Tlsfa; TLSF-a; Tlsfb; TLSF-b; Tpar1
Common Name CXCL12
Gene Symbol CXCL12
Biological Activity ED50 = 50 - 100 ng/mL; determined by the dose-dependent chemotaxis of human PBMCs.
Product Type Protein
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Sequence KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNK
Content And Storage -20°C
Activity ED50 = 50 - 100 ng/mL; determined by the dose-dependent chemotaxis of human PBMCs.
Endotoxin Level <0.1 ng/μg
Shipping Condition Approved for shipment on Wet or Dry Ice
Expression System E. coli
Protein Family Chemokines & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype SDFs (Stromal Cell-Derived Factors)
Research Category Immunology, Stem Cell Research, Cardiovascular Research, Neurobiology
Product Line Gibco
Purity or Quality Grade >98% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.