missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MOS Polyclonal Antibody

Description
MOS Polyclonal Antibody for Western Blot, IHC (P)
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.
Specifications
Specifications
| Antigen | MOS |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | MOS |
| Gene Accession No. | P00540 |
| Gene Alias | C-MOS; c-mos oncogene; c-mos proto-oncogene; MGC119962; MGC119963; Moloney sarcoma oncogene; mos; MOS proto-oncogene, serine/threonine kinase; MOS proto-oncogene, serine/threonine kinase L homeolog; mos.L; mosxe; MSV; oncogene MOS, Moloney murine sarcoma virus; oocyte maturation factor mos; p39; p39-mos; pp39-mos; protein kinase; proto-oncogene c-Mos; Proto-oncogene serine/threonine-protein kinase mos; serine/threonine-protein kinase mos; v-mos Moloney murine sarcoma viral oncogene homolog; v-mos Moloney murine sarcoma viral oncogene homolog L homeolog; XELAEV_18032032mg |
| Gene Symbols | MOS |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?