missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mono-Methyl Phthalate-3,4,5,6-d4, CDN
Specifications
Specifications
| Chemical Name or Material | Monomethyl Phthalate-d4 |
| CAS | 1276197-40-0 |
| Molecular Formula | C9 D4 H4 O4 |
| Quantity | 0.05 g |
| Synonym | Phthalic Acid, Mono-methyl Ester D4 (Phenyl D4), Phthalic-3,4,5,6 D4 Acid Monomethyl Ester, 2,3,4,5-Tetradeuterio-6-methoxycarbonylbenzoic acid |
| InChI Formula | InChI=1 S/C9H8O4/c1-13-9(12)7-5-3-2-4-6(7)8(10)11/h2-5 H,1H3,(H,10,11)/i2D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OC)c(C(=O)O)c1[2 H] |
| IUPAC Name | 2,3,4,5-tetradeuterio-6-methoxycarbonylbenzoic acid |
| Molecular Weight (g/mol) | 184.182 |
| Formula Weight | 184.067 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?