missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Mono-Ball with Valve and Tubing

Product Code. 10083151
Change view
Click to view available options
:
Material: Rubber
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10083151 Material: Rubber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10083151 Supplier Kartell™ Supplier No. 0391000
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Kartell™ Mono-Ball with Valve and Tubing is best suited for pellet pattern burette charging as well as for vaporizers.

  • Suitable for pellet burette charging or for vaporizers
  • 46mm in diameter

Specifications

Color Red
Diameter (Metric) Outer 46 mm
For Use With (Equipment) Pellet Burette Charging
Material Rubber
Includes Valve and Hoses
Type Mono Ball
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.