missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Multiplexed Proteomics™ Phosphoprotein Gel Stain Kits (with Pro-Q™ Diamond and SYPRO™ Ruby Gel Stains)

Product Code. 10104722
Change view
Click to view available options
Quantity:
1 Set, 1 L per Dye
1 Set, 200 mL per Dye
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10104722 1 Set, 200 mL per Dye Each
10788273 1 Set, 1 L per Dye Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10104722 Supplier Invitrogen™ Supplier No. M33306

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Our new Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis.

Our new Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis. This reagent, when combined with our total protein stain, SYPRO Ruby Protein Gel Stain, becomes a powerful combination for quantitative proteome analysis.

Our new Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis. This reagent, when combined with our total protein stain, SYPRO Ruby Protein Gel Stain, becomes a powerful combination for quantitative proteome analysis. This convenient Pro-Q Diamond Phosphoprotein Gel Stain and SYPRO Ruby Protein Gel Stain Pack provides each stain in 1-liter amounts (Cat. No. M-33305) or in 200-mL amounts (Cat. No. M-33306).

Features include:
Sensitive—Pro-Q Diamond Phosphoprotein Gel Stain offers detection level down to 1–16 ng of phosphoprotein per band
Convenient—selectively stain phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis. SYPRO Ruby Gel Stain is subsequently used for total protein staining and simplifies the determination of phosphorylation level
Linear signal—quantify with ease as Pro-Q Diamond Phosphoprotein Gel Stain and SYPRO Ruby Gel Stain exhibit excellent linearity over three orders of magnitude, ideal for quantitation

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Pro-Q Diamond Phosphoprotein Gel Stain, 200 mL
• SYPRO Ruby Protein Gel Stain, 200 mL

Store at room temperature and protect from light.
Description Multiplexed Proteomics™ Phosphoprotein Gel Stain Kit #2
Detection Location In-Gel Detection
Detection Method Fluorescence
Label or Dye SYPRO Ruby, Pro-Q Diamond
Quantity 1 Set, 200 mL per Dye
Shipping Condition Room Temperature
Target Molecule Proteins (Phosphoproteins)
Product Line PRO-Q, SYPRO
Product Type Phosphoprotein Gel Stain Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.