missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP20 Polyclonal Antibody

Description
MMP20 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | MMP20 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | MMP20 |
| Gene Accession No. | O60882 |
| Gene Alias | AI2A2; Enamel metalloproteinase; enamel metalloproteinase precursor; enamelysin; matrix metallo protease; matrix metallopeptidase 20; matrix metallopeptidase 20 (enamelysin); matrix metalloproteinase; matrix metalloproteinase 20; matrix metalloproteinase 20 (enamelysin); matrix metalloproteinase-20; MMP; MMP20; MMP-20; MMPs |
| Gene Symbols | MMP20 |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?