missing translation for 'onlineSavingsMsg'
Learn More

ML-030, MedChemExpress

Product Code. p-200099485
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496507 5 mg 5mg
16453788 10 mg 10mg
16452856 25 mg 25mg
16450167 50 mg 50mg
16414846 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16496507 Supplier Medchem Express Supplier No. HY103050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ML-030 is a potent PDE4 inhibitor, with IC50 of 6.7 nM, 12.9 nM, 48.2 nM, 37.2 nM, 452 nM and 49.2 nM for PDE4A, PDE4A1, PDE4B1, PDE4B2, PDE4C1,and PDE4D2, respectively.

ML-030 is a potent PDE4 inhibitor, with IC50 of 6.7 nM, 12.9 nM, 48.2 nM, 37.2 nM, 452 nM and 49.2 nM for PDE4A, PDE4A1, PDE4B1, PDE4B2, PDE4C1,and PDE4D2, respectively.

Specifications

Chemical Name or Material ML-030
CAS 1013750-77-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H20N4O4S
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (121.22 mM; Need ultrasonic)
SMILES COC1=CC=C(C2=NN3C(SC2)=NN=C3C4=CC(OC)=CC=C4OC)C=C1OC
Molecular Weight (g/mol) 412.46
Formula Weight 412.46
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.