missing translation for 'onlineSavingsMsg'
Learn More

MK-7622, MedChemExpress

Product Code. p-200097290
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496688 10 mg 10mg
16469247 5 mg 5mg
16416846 100 mg 100mg
16402777 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496688 Supplier Medchem Express Supplier No. HY15618

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MK-7622 (M1 receptor modulator) is a muscarinic M1 receptor positive allosteric modulator.

MK-7622 (M1 receptor modulator) is a muscarinic M1 receptor positive allosteric modulator.

Specifications

Chemical Name or Material MK-7622
CAS 1227923-29-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H25N3O2
Quantity 10 mg
Synonym M1 receptor modulator
Solubility Information DMSO : ≥ 100 mg/mL (250.33 mM)
SMILES O=C1N([C@@H]2[C@@H](O)CCCC2)C=NC3=C1C=C(CC4=CC=C(C)N=C4)C5=CC=CC=C53
Molecular Weight (g/mol) 399.48
Formula Weight 399.48
Percent Purity 98.98%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.