missing translation for 'onlineSavingsMsg'
Learn More

MK-3697, MedChemExpress

Product Code. 16497797
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497797 5 mg 5mg
16495078 10 mg 10mg
16461317 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16497797 Supplier Medchem Express Supplier No. HY12301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MK-3697 is an isonicotinamide small molecule, acting as a potent and selective Orexin 2 receptor antagonist with Ki = 0.95 nM.

MK-3697 is an isonicotinamide small molecule, acting as a potent and selective Orexin 2 receptor antagonist with Ki = 0.95 nM.

Specifications

Chemical Name or Material MK-3697
CAS 1224846-01-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H21N5O3S
Quantity 5 mg
Solubility Information DMSO : 22.5 mg/mL (50.28 mM; Need ultrasonic and warming)
SMILES O=C(C1=CC(C2=CC(C)=CN=C2)=NC=C1C3=NC=CS3)NCC4=NC(OC)=C(OC)C=C4
Molecular Weight (g/mol) 447.51
Formula Weight 447.51
Percent Purity 99.46%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.