Learn More
Invitrogen™ Mitochondria Membrane Integrity Antibody Cocktail

Description
Mitochondria Membrane Integrity cocktail of monoclonal antibodies specific to human, bovine, mouse, rat mitochondria membranes. This antibody is validated for use in Western Blot, and can be used to assess mitochondrial membrane integrity during and after purification. Contains: 50 μg of Porin clone #2B12AF12 - Human VDAC1. Concentration 2 μg/mL. 50 μg of Cyclophilin D clone #E11AE12BD4 - Rat CypD/PPIF. Concentration 2 μg/mL. 50 μg of C-III-Core 1 clone # 16D10AD9AH5 - Bovine Heart Complex II/III. Concentration 2 μg/mL. 5 μg of Cytochrome C clone #37BA11 - Human Cytochrome c. Concentration 0.1 μg/mL. 100 μg of C-V-a clone #7H10BD4F9 - Bovine Complex V. Concentration 4 μg/mL. This antibody cocktail (2.37 mg/mL) should be diluted to a final working concentration of 10.2 μg/mL for use in Western blot applications.
Specifications
Specifications
| Antigen | Mitochondria Membrane Integrity |
| Applications | Western Blot |
| Classification | Cocktail |
| Clone | Cocktail |
| Concentration | 2.37 mg/mL |
| Conjugate | Unconjugated |
| Formulation | HEPES buffered saline with 0.02% sodium azide |
| Gene | CYC1 |
| Gene Accession No. | P00125, P08574, P21796, P29117, P30404, P30405, P45879, P62894, P62897, P62898, P99999, Q60932, Q99KR7, Q9D0M3, Q9Z2L0 |
| Gene Alias | 16D10AD9AH5; 20B12AF2; 2610002H19Rik; 37BA11; 7H10BD4F9; AA408921; AL033343; AW457192; BR1-VDAC; brain-derived voltage-dependent anion channel 1; Complex III subunit 4; complex III subunit IV; CYC; Cyc1; cyclophilin 3; cyclophilin D; Cyclophilin F; Cycs; CYCSA; CYP3; CypD; CyP-D; CyP-F; CyP-M; cytc; Cytochrome b-c1 complex subunit 4; cytochrome c; Cytochrome C, expressed in somatic tissues; cytochrome c, somatic; cytochrome c1; Cytochrome c-1; cytochrome c1, heme protein, mitochondrial; E11AE12BD4; HCS; MC3DN6; MGC128356 protein; mitochondrial cyclophilin; mitochondrial Cyclophilin D; mVDAC1; mVDAC5; outer mitochondrial membrane protein porin 1; Peptidyl-prolyl cis-trans isomerase F, mitochondrial; peptidyl-prolyl cis-trans isomerase, mitochondrial; peptidylprolyl isomerase F; peptidylprolyl isomerase F (cyclophilin F); plasmalemmal porin; PORIN; Porin 31HL; porin 31HM; PPIase; PPIase F; PPIF; Rotamase F; rVDAC1; THC4; ubiquinol-cytochrome-c reductase complex cytochrome c1 subunit; Unknown (protein for MGC:134266); UQCR4; VDAC; Vdac1; VDAC-1; Vdac5; VDAC-5; voltage dependent anion channel 1; voltage-dependent anion channel 1; voltage-dependent anion-selective channel protein 1; Voltage-dependent anion-selective channel protein 5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.