missing translation for 'onlineSavingsMsg'
Learn More

Mirogabalin, MedChemExpress

Product Code. 16495936
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495936 100 mg 100mg
16483316 25 mg 25mg
16455278 10 mg 10mg
16431487 50 mg 50mg
16417997 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16495936 Supplier Medchem Express Supplier No. HY12650

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mirogabalin (DS-5565) is a novel, preferentially selective α2δ-1 ligand characterized by high potency and selectivity to the α2δ-1 subunit of voltage-sensitive calcium channel complexes in the CNS.

Mirogabalin (DS-5565) is a novel, preferentially selective α2δ-1 ligand characterized by high potency and selectivity to the α2δ-1 subunit of voltage-sensitive calcium channel complexes in the CNS.

Specifications

Chemical Name or Material Mirogabalin
Physical Form Solid
CAS 1138245-13-2
Color Light Brown
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H19NO2
For Use With (Application) Neuroscience-Neuromodulation
Synonym DS5565
Solubility Information H2O : 7.71 mg/mL (36.84 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES O=C(O)C[C@]1(CN)[C@]2([H])C=C(CC)C[C@]2([H])C1
Molecular Weight (g/mol) 209.28
Formula Weight 209.28
Percent Purity 98.04%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.