missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mirogabalin, MedChemExpress
Description
Mirogabalin (DS-5565) is a novel, preferentially selective α2δ-1 ligand characterized by high potency and selectivity to the α2δ-1 subunit of voltage-sensitive calcium channel complexes in the CNS.
Specifications
Specifications
| Chemical Name or Material | Mirogabalin |
| Physical Form | Solid |
| CAS | 1138245-13-2 |
| Color | Light Brown |
| Quantity | 25 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C12H19NO2 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Synonym | DS5565 |
| Solubility Information | H2O : 7.71 mg/mL (36.84 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?