missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Water Testing Minikit

Product Code. 10024230
Change view
Click to view available options
Range:
0 to 500 mg/L
0.75 to 10% acid
10 to 500 mg/L
2 to 50 mg/L
20 to 200 mg/L
20 to 500 mg/L
5 to 5000 mg/L
70 to 1500 mg/L
Analytes:
Alkalinity Total M
Alkalinity-P, CaC03
Chloride
Cleaning Acid Strength
Cyanuric Acid
Nitrite
Sulphate (low range) Na2SO4
Sulphite (high range)
Sulphite (low range)
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Analytes unitSize
10024230 0 to 500 mg/L Sulphite (high range) Each
10268420 5 to 5000 mg/L Chloride Each
10248520 70 to 1500 mg/L Nitrite Each
10511931 2 to 50 mg/L Sulphite (low range) Each
10616151 20 to 200 mg/L Cyanuric Acid Each
10501931 20 to 200 mg/L Sulphate (low range) Na2SO4 Each
13151701 0.75 to 10% acid Cleaning Acid Strength Each
10014480 10 to 500 mg/L Alkalinity Total M Each
10228330 20 to 500 mg/L Alkalinity-P, CaC03 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 10024230 Supplier Lovibond™ Supplier No. 414350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lovibond™ Water Testing Minikit is easy to use and provides highly accurate results.

  • Easy operation and exact reagent dosing
  • High accuracy
  • Foil-wrapped Lovibond™ tablet reagents with a minimum guaranteed shelf life of 5 years
  • Unrestricted shipment
  • Safe storage

Specifications

Analytes Sulphite (high range)
Includes Flip top case containing a shaker tube, rapid dissolving tablet reagents and instructions.
Quantity 1 Each
Range 0 to 500 mg/L
Sufficient For Approximately 40 Tests
Test Method Tablet count
Type Minikit water testing kit
compliance-icons
Product Identifier
  • MINIKIT TEST SULFITE HIGH AF435
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: sulfamic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.