missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Water Testing Minikit
Description
Lovibond™ Water Testing Minikit is easy to use and provides highly accurate results.
- Easy operation and exact reagent dosing
- High accuracy
- Foil-wrapped Lovibond™ tablet reagents with a minimum guaranteed shelf life of 5 years
- Unrestricted shipment
- Safe storage
Specifications
Specifications
| Analytes | Chloride |
| Includes | Flip top case containing a shaker tube, rapid dissolving tablet reagents and instructions. |
| Quantity | 1 Pack |
| Range | 5 to 5000 mg/L |
| Sufficient For | Approximately 40 Tests |
| Test Method | Tablet count |
| Type | Minikit water testing kit |
Safety and Handling
Product Identifier
- MINIKIT TEST CHLORIDE SALINITY AF418
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360-May damage fertility or the unborn child.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- MIXTURE LIST-Contain: boric acid, potassium chromate, silver nitrate, potassium dichromate
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?